Learn More
Sino Biological Recombinant Mouse MIF Protein
Supplier:  Sino Biological 50066MNAE50
Encompass_Preferred
A DNA sequence encoding the mouse MIF (P34884) precursor (Pro 2-Ala 115) was expressed, with an initial Met at the N-terminus.
Catalog No. 50-162-1275