Learn More
Sino Biological Recombinant Mouse 4-1BB Ligand/TNFSF9 Protein (His Tag)
Supplier:  Sino Biological 50067M07H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Arg 104-Glu 309) of mouse TNFSF9 (NP_033430.1) precursor was expressed with a N-terminal polyhistidine tag.
Catalog No. 50-162-1278