Learn More
Sino Biological Recombinant Mouse B7-2/CD86 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50068M08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Glu 245) of mouse CD86 (NP_062261.3) precursor was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1281