Learn More
Sino Biological Recombinant Mouse GRO Beta/CXCL2 Protein (His Tag)
Supplier:  Sino Biological 50070M08Y10
Encompass_Preferred
A DNA sequence encoding the mouse Cxcl2 (NP_033166.1) (Ala28-Asn100) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1286