Learn More
Sino Biological Recombinant Mouse BMPR1A/ALK-3 Protein (His Tag)
Supplier:  Sino Biological 50078M08H200
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Arg 152) of mouse ALK3 (NP_033888.2) precursor was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1307