Learn More
Sino Biological Recombinant Mouse ASGR1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 50083M07H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Ser 60-Asn 284) of mouse ASGPR1 (NP_033844.1) was expressed with a N-terminal polyhistidine tag.
Catalog No. 50-162-1310