Learn More
Sino Biological Recombinant Mouse IL-7R alpha/CD127 Protein (His Tag)
Supplier:  Sino Biological 50090M08H100
Encompass_Preferred
A DNA sequence encoding the mouse IL7Rα (NP_032398.3) extracellular domain (Met 1-Asp 239) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-1330