Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 500g, Bis(pinacolato)diboron, 99%, [73183-34-3], D6878 253.94 C12H24B2O4

Catalog No. 5036756
Encompass_Preferred

Compound: Bis(pinacolato)diboron

CAS#: 73183-34-3

MDL#: MFCD00799570

Background: The most commonly used diboron reagent in organic synthesis due to its high stability in air and moisture. It is used for Ir- or other transition metal-catalyzed C-H borylation of arenes, heteroarenes, and alkylarenes, and for the diborylation of alkynes and alkenes to generate aryl-, heteroaryl-, and alkylboron compounds.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-367-56 Supplier Frontier Specialty Chemicals Supplier No. D6878500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Bis(pinacolato)diboron

CAS#: 73183-34-3

MDL#: MFCD00799570

Background: The most commonly used diboron reagent in organic synthesis due to its high stability in air and moisture. It is used for Ir- or other transition metal-catalyzed C-H borylation of arenes, heteroarenes, and alkylarenes, and for the diborylation of alkynes and alkenes to generate aryl-, heteroaryl-, and alkylboron compounds.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.