Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 500mg, 1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride, 95%, [250285-32-6], B13197 425.049 C27H37ClN2

Catalog No. 5034803
Encompass_Preferred

Compound: 1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride

CAS#: 250285-32-6

MDL#: not listed

Background: An N-heterocyclic carbene (NHC) precursor salt, known as IPr·HCl, widely used to generate bulky, electron-rich NHC ligands for transition metal catalysis. It is a key component in many palladium- and ruthenium-catalyzed cross-coupling and olefin metathesis catalyst systems.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-348-03 Supplier Frontier Specialty Chemicals Supplier No. B13197500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride

CAS#: 250285-32-6

MDL#: not listed

Background: An N-heterocyclic carbene (NHC) precursor salt, known as IPr·HCl, widely used to generate bulky, electron-rich NHC ligands for transition metal catalysis. It is a key component in many palladium- and ruthenium-catalyzed cross-coupling and olefin metathesis catalyst systems.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.