Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 500mg, 2,2,2-Trifluoroacetophenone-3-boronic acid pinacol ester, 97%, [631909-42-7], T13186 300.081 C14H16BF3O3

Catalog No. 5039297
Encompass_Preferred

Compound: 2,2,2-Trifluoroacetophenone-3-boronic acid pinacol ester

CAS#: 631909-42-7

MDL#: not listed

Background: A trifluoromethyl ketone-substituted phenylboronic ester used as a Suzuki-Miyaura coupling partner in pharmaceutical research. The trifluoromethyl ketone group is a reactive electrophile useful for further derivatization after coupling.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-392-97 Supplier Frontier Specialty Chemicals Supplier No. T13186500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2,2,2-Trifluoroacetophenone-3-boronic acid pinacol ester

CAS#: 631909-42-7

MDL#: not listed

Background: A trifluoromethyl ketone-substituted phenylboronic ester used as a Suzuki-Miyaura coupling partner in pharmaceutical research. The trifluoromethyl ketone group is a reactive electrophile useful for further derivatization after coupling.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.