Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 500mg, Fe(III) meso-Tetraphenylporphine Chloride (contains 1-3% chlorin), >95%, [16456-81-8], T40253 704.018 C44H28ClFeN4

Catalog No. 5039313
Encompass_Preferred

Compound: Fe(III) meso-Tetraphenylporphine Chloride (contains 1-3% chlorin)

CAS#: 16456-81-8

MDL#: not listed

Background: A specialty chemical manufactured by Frontier Specialty Chemicals useful for numerous catalytic organic transformations. It catalyzes a highly E-selective olefination of aldehydes with ethyl diazoacetate, and selective Baeyer-Villiger oxidation of cycloketones to lactones with molecular oxygen.

Storage: Store at room temperature, protected from light.

Catalog No. 50-393-13 Supplier Frontier Specialty Chemicals Supplier No. T40253500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Fe(III) meso-Tetraphenylporphine Chloride (contains 1-3% chlorin)

CAS#: 16456-81-8

MDL#: not listed

Background: A specialty chemical manufactured by Frontier Specialty Chemicals useful for numerous catalytic organic transformations. It catalyzes a highly E-selective olefination of aldehydes with ethyl diazoacetate, and selective Baeyer-Villiger oxidation of cycloketones to lactones with molecular oxygen.

Storage: Store at room temperature, protected from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.