Learn More
Avanti Polar Lipids 500MG LYSO DDPG (16:0 LYSO PG)
Supplier:  Avanti Polar Lipids 858122P500MG
Encompass
1-palmitoyl-2-hydroxy-sn-glycero-3-phospho-(1'-rac-glycerol) (sodium salt). 326495-22-1
Catalog No. NC0212321