Learn More
Avanti Polar Lipids 1,2-dioleoyl-3-trimethylammonium-propane (chloride salt), 132172-61-3, MFCD01310872, 500mg
Catalog No. NC1120152
Encompass
Molecular Formula: C42H80NO4Cl, Formula Weight: 698.542, Purity >99%. Cationic liposome-forming compound for transfection of DNA, RNA and other negatively charged molecules into eukaryotic cells. Mixing DOTAP with DNA results in spontaneously formed stable complexes that can be directly added to cell culture medium. These complexes fuse with the cell membrane and release DNA into the cytoplasm. The cells are transfected efficiently without cytotoxic effects.
Catalog No. NC1120152 Supplier Avanti Polar Lipids Supplier No. 890890P500MG
Molecular Formula: C42H80NO4Cl, Formula Weight: 698.542, Purity >99%. Cationic liposome-forming compound for transfection of DNA, RNA and other negatively charged molecules into eukaryotic cells. Mixing DOTAP with DNA results in spontaneously formed stable complexes that can be directly added to cell culture medium. These complexes fuse with the cell membrane and release DNA into the cytoplasm. The cells are transfected efficiently without cytotoxic effects.