Learn More
Inorganic Ventures 10 000µg/mL SULFATE 500mL
SDP
Catalog No. NC0813251
Encompass_Preferred
Matrix: Water (H2O), Starting Material: K2SO4, Analyte: SO4-2 10000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. NC0813251 Supplier Inorganic Ventures Supplier No. ICSO410500ML
Matrix: Water (H2O), Starting Material: K2SO4, Analyte: SO4-2 10000 ug/mL.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search