Learn More
Ace Glass, Inc. 500mL 24/25 separatory funnel with polyethylene stopper
SDP
Catalog No. 50974691
Encompass_Preferred
500ml separatory funnel with polyethylene stopper. Outer ground joint at top is a 24/25 Rodaviss joint. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.
Catalog No. 50-974-691 Supplier Ace Glass, Inc. Supplier No. 723115
Add to Cart
Add to Cart
500ml separatory funnel with polyethylene stopper. Outer ground joint at top is a 24/25 Rodaviss joint. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.