Learn More
Ace Glass, Inc. 500mL graduated cylindrical funnel, 24/40 top, 24/40 bottom
SDP
Catalog No. 50974738
Encompass_Preferred
500mL graduated cylindrical funnel with 24/40 joints at top and bottom. Features include drip stem, and double scale graduations calibrated from stopcock. Drip stem does not extend beyond the joint, thereby eliminating a possible source of breakage. Includes PTFE stopcock plug.
Catalog No. 50-974-738 Supplier Ace Glass, Inc. Supplier No. 726864
Add to Cart
Add to Cart
500mL graduated cylindrical funnel with 24/40 joints at top and bottom. Features include drip stem, and double scale graduations calibrated from stopcock. Drip stem does not extend beyond the joint, thereby eliminating a possible source of breakage. Includes PTFE stopcock plug.