Learn More
Ace Glass, Inc. 500mL epoxy coated flash chromatography reservoir with 35/20upper joint and 35/20 lower joint.
SDP
Catalog No. 50972489
Encompass_Preferred
500mL epoxy coated flash chromatography reservoir with 35/20 upper and lower joints. Reservoir connects between the column and flow control adapter.
Catalog No. 50-972-489 Supplier Ace Glass, Inc. Supplier No. 587242
500mL epoxy coated flash chromatography reservoir with 35/20 upper and lower joints. Reservoir connects between the column and flow control adapter.