Learn More
Chemglass Life Sciences Single Port Vent Assembly, GL-45, Universal 316L Electro Polished Stainless Steel, 6.35mm/0.25" OD Hose Barb, Complete with Gasket and Cap
SDP
Catalog No. 501294531
Encompass
Universal vent port assembly is constructed of 316L electro polished stainless steel.These port assemblies fit all types of spinner flasks, media bottles and vessels.Configurations consist of a single vent tube, vent and short fill tubes, or angled vent and fill tubes.Supplied complete with a GL-32 or GL-45 open top cap and silicone gasket.
Catalog No. 50-129-4531 Supplier Chemglass Life Sciences Supplier No. CLS142910
Universal vent port assembly is constructed of 316L electro polished stainless steel.These port assemblies fit all types of spinner flasks, media bottles and vessels.Configurations consist of a single vent tube, vent and short fill tubes, or angled vent and fill tubes.Supplied complete with a GL-32 or GL-45 open top cap and silicone gasket.