Learn More
Chemglass Life Sciences 500mL Addition Funnel, 24/40 Joint Size, 4mm Stpk, 435mm OAH
SDP
Catalog No. 50873263
Encompass_Preferred
500mL Addition Funnel, 24/40 Joint Size, 4mm Stopcock, 435mm OAH. Cylindrical shape addition funnel with PTFE stopcocks. For replacement stoppers see CG-3000.
Catalog No. 50-873-263 Supplier Chemglass Life Sciences Supplier No. CG170804
500mL Addition Funnel, 24/40 Joint Size, 4mm Stopcock, 435mm OAH. Cylindrical shape addition funnel with PTFE stopcocks. For replacement stoppers see CG-3000.