Learn More
Chemglass Life Sciences 500mL Addition Funnel, 24/40 Joint Size, 4mm Glass Stpk, 435mm OAH
SDP
Catalog No. CG170004
Encompass_Preferred
500mL Addition Funnel, 24/40 Joint Size, 4mm Glass Stopcock, 435mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and glass stopcock. For replacement stoppers see CG-3000.
Catalog No. CG170004 Supplier Chemglass Life Sciences Supplier No. CG170004
500mL Addition Funnel, 24/40 Joint Size, 4mm Glass Stopcock, 435mm OAH. Cylindrical style addition funnel with a standard taper top outer joint and lower inner joint, both being the same size. With pressure equalizing arm and glass stopcock. For replacement stoppers see CG-3000.