Learn More
Chemglass Life Sciences Bottle Only, 500mL Gas Washing
SDP
Catalog No. CG111209
Encompass_Preferred
Bottle Only, 500mL Gas Washing. Replacement part for item CG-1112-03 Tall form gas washing bottle having a standard taper 34/28 top joint, hex base and plain tube for gas dispersion.
Catalog No. CG111209 Supplier Chemglass Life Sciences Supplier No. CG111209
Bottle Only, 500mL Gas Washing. Replacement part for item CG-1112-03 Tall form gas washing bottle having a standard taper 34/28 top joint, hex base and plain tube for gas dispersion.