Learn More
Ace Glass, Inc. Trap, bump, 500mL, 24/40 top, 24/40 bottom
SDP
Catalog No. NC0622319
Encompass_Preferred
500mL bump trap for use between the rotary evaporators steam tube and evaporator flask, preventing fluid from being drawn into the condenser. Trap has 24/40 top and bottom joints.
Catalog No. NC0622319 Supplier Ace Glass, Inc. Supplier No. 670424
500mL bump trap for use between the rotary evaporators steam tube and evaporator flask, preventing fluid from being drawn into the condenser. Trap has 24/40 top and bottom joints.