Learn More
NEST SCIENTIFIC USA 500ML CENTR TUBES W/ PREAS 4CS
Catalog No. NC3311163
Encompass_Preferred
500mL Centrifuge Tube with Bi-directional Transfer Cap with Silicon Tube (50cm 1/8" ID 1/4" OD), Male Luer Lock Connector with Luer Cap, Vent Filter (0.22 μm Φ24mm), Sterile, 1/pk, 4/cs
Catalog No. NC3311163 Supplier NEST SCIENTIFIC USA Supplier No. C61022EFB050A
Add to Cart
Add to Cart
500mL Centrifuge Tube with Bi-directional Transfer Cap with Silicon Tube (50cm 1/8" ID 1/4" OD), Male Luer Lock Connector with Luer Cap, Vent Filter (0.22 μm Φ24mm), Sterile, 1/pk, 4/cs