Learn More
Bellco Glass, Inc. 500mL Centrifuge Bot Polycarbo
SDP
Catalog No. 502329314
Encompass_Preferred
Centrifuge Bottles have a rigid body for low to moderate speed centrifugation of biological and chemical samples. High quality polycarbonate and polypropylene have excellent chemical resistance. Centrifuge Bottles are reusable and autoclavable.Polycarbonate Seal cap has extended lip to facilitate removal from rotor and gripping ridges for easy opening and sealing Bottles include molded graduation lines Bottles should be filled to 80 of total capacity for maximum resultsSpecifications Size 500mL Pack Description 4/Bag Max Force RCF g 13 850 Case Quantity 4
Catalog No. 50-232-9314 Supplier Bellco Glass, Inc. Supplier No. 229469000
Centrifuge Bottles have a rigid body for low to moderate speed centrifugation of biological and chemical samples. High quality polycarbonate and polypropylene have excellent chemical resistance. Centrifuge Bottles are reusable and autoclavable.Polycarbonate Seal cap has extended lip to facilitate removal from rotor and gripping ridges for easy opening and sealing Bottles include molded graduation lines Bottles should be filled to 80 of total capacity for maximum resultsSpecifications Size 500mL Pack Description 4/Bag Max Force RCF g 13 850 Case Quantity 4