Learn More
Bellco Glass, Inc. 500mL Centrifuge Bottle
SDP
Catalog No. 502330865
Encompass_Preferred
DescriptionCentrifuge Bottles have a rigid body for low to moderate speed centrifugation of biological and chemical samplesManufactured from high quality polypropylene for excellent chemical resistanceBottle must be filled 80% of total capacity to obtain maximum resultsCentrifuge Bottles are reusable and autoclavablePackaged non-sterile, 2 per bag Pack Description2/BagCase Quantity2SterileNoAutoclavableYesPrimary ColorNaturalPrimary MaterialPolypropyleneMaximum Volume500mLGraduatedNoMax Force (RCF)g13500
Catalog No. 50-233-0865 Supplier Bellco Glass, Inc. Supplier No. 229468000
DescriptionCentrifuge Bottles have a rigid body for low to moderate speed centrifugation of biological and chemical samplesManufactured from high quality polypropylene for excellent chemical resistanceBottle must be filled 80% of total capacity to obtain maximum resultsCentrifuge Bottles are reusable and autoclavablePackaged non-sterile, 2 per bag Pack Description2/BagCase Quantity2SterileNoAutoclavableYesPrimary ColorNaturalPrimary MaterialPolypropyleneMaximum Volume500mLGraduatedNoMax Force (RCF)g13500