Learn More
CELLTREAT Scientific Products 500mL Centrifuge Bottle, PP, Non-sterile
SDP
Catalog No. NC3433418
Encompass_Preferred
"Translucent and provides chemical resistivity needed for many applicationsBottle should be filled 75% of total capacity to obtain maximum resultsCentrifuge Bottles have a rigid body for low to moderate speed centrifugation of biological and chemical samplesHigh quality polycarbonate and polypropylene have excellent chemical resistanceReusable and autoclavableUse of a rotor cushion is advised to support bottles during centrifugationPackaged non-sterile in re-sealable bags"
Catalog No. NC3433418 Supplier CELLTREAT Scientific Products Supplier No. 229468
"Translucent and provides chemical resistivity needed for many applicationsBottle should be filled 75% of total capacity to obtain maximum resultsCentrifuge Bottles have a rigid body for low to moderate speed centrifugation of biological and chemical samplesHigh quality polycarbonate and polypropylene have excellent chemical resistanceReusable and autoclavableUse of a rotor cushion is advised to support bottles during centrifugationPackaged non-sterile in re-sealable bags"