Learn More
Alkali Scientific 500mL Centrifuge Tube with Bi-
SDP
Supplier:  Alkali Scientific BDTC61022
Encompass_Preferred
The pre-assembled Closed System of Centrifuge Tube is convenient for aseptic sampling and transfer during biological production procedures Each is double-packed and equipped with a seal cap which can be used to substitute the pipes in case of subsequent centrifugation and analysis The pre-assembled Closed System of Centrifuge Tube is convenient for customers in biomedical/biopharmaceutical/vaccine production industries to aseptically sample and transfer the tissue and cell culture/ vaccine solutions which can effectively save the assembling and sampling time and lower the cost The pre-assembled Closed System is ready to use at hand The closed liquid transfer avoids the contamination risk of open transfer and eliminates the necessity of subsequent cleaning validation The pipeline material meets the requirements of industrial pharmaceutical enterprises and is connected to male taper at the connector which prevents accidental disconnection and offers convenience The double aseptic p
Catalog No. 50-302-6540