Learn More
Ace Glass, Inc. 500ML DIST APPARATUS C 500Ml Dist Apparatus C
SDP
Catalog No. 50978823
Encompass_Preferred
Complete distillation apparatus generally used for the determination of ammonia in water. Joints are standard taper 24/40.
Catalog No. 50-978-823 Supplier Ace Glass, Inc. Supplier No. 655310
Complete distillation apparatus generally used for the determination of ammonia in water. Joints are standard taper 24/40.