Learn More
Chemglass Life Sciences 500mL Glassblowers Erlenmeyer Blank
SDP
Catalog No. CG62006
Encompass_Preferred
500mL Glassblowers Erlenmeyer Blank. Blanks for fabrication of Erlenmeyer shaped flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.
Catalog No. CG62006 Supplier Chemglass Life Sciences Supplier No. CG62006
500mL Glassblowers Erlenmeyer Blank. Blanks for fabrication of Erlenmeyer shaped flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.