Learn More
Chemglass Life Sciences Flask, Erlenmeyer, 500mL, PETG, Flat Base, with Plug Seal Cap, Sterile, 1/pk, 12/cs
SDP
Catalog No. 501897648
Encompass_Preferred
PETG flask with HDPE cap.Sterilized by E-Beam.Nonpyrogenic.Packaged in sterile bags.
Catalog No. 50-189-7648 Supplier Chemglass Life Sciences Supplier No. CGN2092P500
PETG flask with HDPE cap.Sterilized by E-Beam.Nonpyrogenic.Packaged in sterile bags.