Learn More
Chemglass Life Sciences Glas-Col Mantle Extension Support for 500mL Series O Mantle, 5 7/8in ID
SDP
Catalog No. CG1001004
Encompass_Preferred
Glas-Col Mantle Extension Support for 500mL Series O Mantle, 5 7/8in ID. This lattice mount support holds spherical and hemispherical Glas-Col Series O mantles up to 5-liter capacity. The steel support, with its fabric strap basket, is strong, handy and durable. Attaches easily to stable ring stands or distillation racks. Supports are painted gray. This mantle support is made from 3/8in diameter rod.
Catalog No. CG1001004 Supplier Chemglass Life Sciences Supplier No. CG1001004
Glas-Col Mantle Extension Support for 500mL Series O Mantle, 5 7/8in ID. This lattice mount support holds spherical and hemispherical Glas-Col Series O mantles up to 5-liter capacity. The steel support, with its fabric strap basket, is strong, handy and durable. Attaches easily to stable ring stands or distillation racks. Supports are painted gray. This mantle support is made from 3/8in diameter rod.