Learn More
Ace Glass, Inc. Outer extraction chamber, 500mL
SDP
Catalog No. 50973447
Encompass_Preferred
500mL outer extraction chamber designed for the extracting of liquids with ether. The dispersion of the ether is accomplished with the use of a 70-100micron porosity ACE fritted disc, which is attached to the base of the funnel tube.
Catalog No. 50-973-447 Supplier Ace Glass, Inc. Supplier No. 684019
500mL outer extraction chamber designed for the extracting of liquids with ether. The dispersion of the ether is accomplished with the use of a 70-100micron porosity ACE fritted disc, which is attached to the base of the funnel tube.