Learn More
Chemglass Life Sciences 500mL Glassblowers Flat Bottom Flask Blank
SDP
Catalog No. CG61904
Encompass_Preferred
500mL Glassblowers Flat Bottom Flask Blank. Blanks for fabrication of single or multi-neck flat bottom flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.
Catalog No. CG61904 Supplier Chemglass Life Sciences Supplier No. CG61904
500mL Glassblowers Flat Bottom Flask Blank. Blanks for fabrication of single or multi-neck flat bottom flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.