Learn More
CELLTREAT Scientific Products 500mL Filter System, 0.22um PES Filter, 90mm Diameter, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229707
Encompass_Preferred
CELLTREAT Scientific Products 500mL, 0.22um PES Filter System. Easy-to-read molded graduations marks on bottle top filter. Bold easy-to-read printed membrane and pore size for easy identification. Curved bottle design for easy handling. Printed lot number and expiration date on every bottle top. Improved filter support provides faster flow rates. Angled hose barb adapter fits three size hose diameters and prevents the filter from tipping. Polyethersulfone (PES) membrane is for high flow and low binding. 0.22um PES membrane is the first choice for sterile filtration of cell culture media. Filter Systems include bottle top filter and solution bottle with cap. Membrane diameter 90mm. Every case is printed with the lot number for quality traceability. Packaged sterile, individually bagged, 12/case.
Catalog No. 50-202-045