Learn More
Chemglass Life Sciences 500mL Filter System, PES, 0.22µm
SDP
Supplier:  Chemglass Life Sciences CGE2009007
Encompass
Disposable filter systems are useful for large volume sample separation and sterilization for tissue culture media and other biological buffers. PES membrane filters with graduated filter tops made with polystyrene, and polyethylene necks with polystyrene receiving bottles. A separate sterile polyethylene cap is included.
Catalog No. 50-189-7580