Learn More
Bellco Glass, Inc. 500mL Filter System PES
SDP
Supplier:  Bellco Glass, Inc. 229707000
Encompass_Preferred
Filter Systems include bottle top filter and solution bottle with cap Bottle Top Filter available separately .Specifications Easytoread molded graduations marks on bottle top filter Bold easytoread printed membrane and pore size for easy identification Curved bottle design for easy handling Printed lot number and expiration date on every bottle top Improved filter support provides faster flow rates Angled hose barb adapter fits three size hose diameters and prevents the filter from tipping Polyethersulfone PES membrane is for high flow and low binding 0.10µm PES membrane ideal for mycoplasma removal 0.22µm PES membrane is the first choice for sterile filtration of cell culture media 0.45µm PES membrane ideal for solution clarification Ca
Catalog No. 50-232-9491