Learn More
Chemglass Life Sciences 500mL Glassblowers RBF Blank
SDP
Catalog No. 50873565
Encompass_Preferred
500mL Glassblowers RBF Blank. Blanks for fabrication of single or multi-neck round bottom flasks. NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.
Catalog No. 50-873-565 Supplier Chemglass Life Sciences Supplier No. CG61809
500mL Glassblowers RBF Blank. Blanks for fabrication of single or multi-neck round bottom flasks. NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.