Learn More
Chemglass Life Sciences 500mL Glassblowers Heavy Wall RBF Blank
SDP
Catalog No. 501531798
Encompass_Preferred
500mL Glassblowers Heavy Wall Round Bottomed Flask Blank. Heavy Wall blanks are 30% heavier than Chemglass's standard CG-618 series. Blanks are used in the fabrication of single or multi-neck round bottom flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.
Catalog No. 50-153-1798 Supplier Chemglass Life Sciences Supplier No. CG61809HW
500mL Glassblowers Heavy Wall Round Bottomed Flask Blank. Heavy Wall blanks are 30% heavier than Chemglass's standard CG-618 series. Blanks are used in the fabrication of single or multi-neck round bottom flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.