Learn More
Chemglass Life Sciences Chemglass 500 mL Sterile Polycarbonate Erlenmeyer Flask with Blue Duo Cap, Case of 12 (CLS-2092-500S)
SDP
Catalog No. NC1947075
Encompass_Preferred
Chemglass Life Sciences sterile 500 mL Erlenmeyer flask made from polycarbonate and supplied with a blue two-piece cap. The flask is designed for mixing, shaking, and short-term storage of biological fluids and is supplied sterile in cases of 12.
  • 500 mL capacity suitable for small-scale culture and mixing.
  • Polycarbonate construction for clarity and impact resistance.
  • 38-430 thread with a blue two-piece cap for secure closure.
  • Supplied sterile and non-pyrogenic for bioprocess applications.
  • Packaged as a case of 12 for lab convenience.
Catalog No. NC1947075 Supplier Chemglass Life Sciences Supplier No. CLS2092500S
Chemglass Life Sciences sterile 500 mL Erlenmeyer flask made from polycarbonate and supplied with a blue two-piece cap. The flask is designed for mixing, shaking, and short-term storage of biological fluids and is supplied sterile in cases of 12.
  • 500 mL capacity suitable for small-scale culture and mixing.
  • Polycarbonate construction for clarity and impact resistance.
  • 38-430 thread with a blue two-piece cap for secure closure.
  • Supplied sterile and non-pyrogenic for bioprocess applications.
  • Packaged as a case of 12 for lab convenience.