Learn More
Chemglass Life Sciences 500mL Erlenmeyer Flask, 29/42 Outer Joint, Graduated
SDP
Catalog No. CG154210
Encompass_Preferred
500mL Erlenmeyer Flask, 29/42 Outer Joint, Graduated. Erlenmeyer flask with a standard taper outer joint. For stoppers see CG-3000.
Catalog No. CG154210 Supplier Chemglass Life Sciences Supplier No. CG154210
500mL Erlenmeyer Flask, 29/42 Outer Joint, Graduated. Erlenmeyer flask with a standard taper outer joint. For stoppers see CG-3000.