Learn More
Ace Glass, Inc. Combo mantle, 3 position, low temperature, fits 500mL flatbottom flasks, max diameter 4.06 inches, 120v 180 watts per position, order code (-61) or (-71) power supply separately
SDP
Catalog No. 50983292
Encompass
Combo mantle, 3 position, low temperature, fits 500mL flatbottom flasks, max diameter 4.06 inches, 120v 180 watts per position, order code (-61) or (-71) power supply separately
Catalog No. 50-983-292 Supplier Ace Glass, Inc. Supplier No. 12061102
Combo mantle, 3 position, low temperature, fits 500mL flatbottom flasks, max diameter 4.06 inches, 120v 180 watts per position, order code (-61) or (-71) power supply separately