Learn More
Ace Glass, Inc. Combo mantle, 3 position, low temperature, fits 500mL flatbottom flasks, max diameter 4.06 inches, 240v 180 watts per position, order code (-63) or (-73) power supply separately
SDP
Catalog No. 50977364
Encompass
Combo mantle, 3 position, low temperature, fits 500mL flatbottom flasks, max diameter 4.06 inches, 240v 180 watts per position, order code (-63) or (-73) power supply separately
Catalog No. 50-977-364 Supplier Ace Glass, Inc. Supplier No. 12061123
Combo mantle, 3 position, low temperature, fits 500mL flatbottom flasks, max diameter 4.06 inches, 240v 180 watts per position, order code (-63) or (-73) power supply separately