Learn More
Ace Glass, Inc. 500ML FUNNEL 500ML FUNNEL
SDP
Catalog No. 50974731
Encompass_Preferred
500mL graduated cylindrical addition funnel with a 24/40 standard taper stopper joint. Stopcock and Stopper not supplied.
Catalog No. 50-974-731 Supplier Ace Glass, Inc. Supplier No. 726254
500mL graduated cylindrical addition funnel with a 24/40 standard taper stopper joint. Stopcock and Stopper not supplied.