Learn More
Chemglass Life Sciences 500mL Funnel Only, Coarse Frit, #50 O-Ring Joint, 70 O.D., 75 Pinchclamp
SDP
Catalog No. CG140514
Encompass_Preferred
500mL Funnel Only, Coarse Frit, #50 O-Ring Joint, 70 O.D., 75 Pinchclamp. For positive pressure filtration of materials up to 15 psi. Connection between the funnel and adapter is made by an o-ring joint which permits a larger, grease free opening for sample retrieval. The 24/40 lower inner standard taper joint has an outlet for connection to a vacuum source.
Catalog No. CG140514 Supplier Chemglass Life Sciences Supplier No. CG140514
500mL Funnel Only, Coarse Frit, #50 O-Ring Joint, 70 O.D., 75 Pinchclamp. For positive pressure filtration of materials up to 15 psi. Connection between the funnel and adapter is made by an o-ring joint which permits a larger, grease free opening for sample retrieval. The 24/40 lower inner standard taper joint has an outlet for connection to a vacuum source.