Learn More
Chemglass Life Sciences Complete 500mL Bottle, Gas Washing, Tall Form, Coarse Frit, 380mm Approx. OAH
SDP
Catalog No. 50872940
Encompass_Preferred
Complete 500mL Bottle, Gas Washing, Tall Form, Coarse Frit, 380mm Approx. OAH. Tall form gas washing bottle having a standard taper 34/28 top joint, hex base and coarse fritted tube for gas dispersion. Stopper is held in place with two CG-110-02, 18mm stainless steel springs. Springs are supplied when bottle is ordered complete.
Catalog No. 50-872-940 Supplier Chemglass Life Sciences Supplier No. CG111415
Complete 500mL Bottle, Gas Washing, Tall Form, Coarse Frit, 380mm Approx. OAH. Tall form gas washing bottle having a standard taper 34/28 top joint, hex base and coarse fritted tube for gas dispersion. Stopper is held in place with two CG-110-02, 18mm stainless steel springs. Springs are supplied when bottle is ordered complete.