Learn More
Chemglass Life Sciences 500mL Addition Funnel, Graduated, 29/42 Joint Size, 4mm PTFE Stpk, 445mm OAH
SDP
Catalog No. CG171205
Encompass_Preferred
500mL Addition Funnel, Graduated, 29/42 Joint Size, 4mm PTFE Stopcock, 445mm OAH. Cylindrical style addition funnel similar to CG-1710, but with a PTFE stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.
Catalog No. CG171205 Supplier Chemglass Life Sciences Supplier No. CG171205
500mL Addition Funnel, Graduated, 29/42 Joint Size, 4mm PTFE Stopcock, 445mm OAH. Cylindrical style addition funnel similar to CG-1710, but with a PTFE stopcock. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper. For replacement stoppers see CG-3000.