Learn More
Chemglass Life Sciences 500mL Kuderna-Danish Flask without Hooks
SDP
Catalog No. CG130813
Encompass_Preferred
500mL Kuderna-Danish Flask without Hooks. Lower body of flask has a hemispherical shape to afford continuous flow of heavier fractions to concentrator tube during reflux. Top joint is a 24/40 outer while the lower joint is a 19/22 inner. For the item that has hooks on the lower 19/22 joint, see CG130803.
Catalog No. CG130813 Supplier Chemglass Life Sciences Supplier No. CG130813
500mL Kuderna-Danish Flask without Hooks. Lower body of flask has a hemispherical shape to afford continuous flow of heavier fractions to concentrator tube during reflux. Top joint is a 24/40 outer while the lower joint is a 19/22 inner. For the item that has hooks on the lower 19/22 joint, see CG130803.