Learn More
Foxx Life Sciences 500mL Narrow Mouth LDPE Reagent Bottles, 48CS
Catalog No. 5000846679
Encompass_Preferred
EZLabpure™  500mL LDPE Reagent Bottles Non Sterile provide safe, leak-proof containment for lab chemicals, solutions & powders. Compliant with USP Class VI & US FDA 21 CFR standards, these shatterproof bottles offer a safer, high-impact alternative to glass in high-handling environments. The Narrow Mouth LDPE Reagent Bottle design ensures precision dispensing, controlled pouring, and minimal spillage. LDPE chemical resistant reagent bottles offer excel chemical resistance and have high impact strength
Features
LDPE storage reagent bottles are flexible and squeezable for easy dispensing.
LDPE storage bottles are lightweight and crack-resistant.
LDPE Bottles with low Liquid Retention
Leak-proof Linerless screw caps for secure sealing.
Catalog No. 50-008-46679 Supplier Foxx Life Sciences Supplier No. 7641G21FLS
EZLabpure™  500mL LDPE Reagent Bottles Non Sterile provide safe, leak-proof containment for lab chemicals, solutions & powders. Compliant with USP Class VI & US FDA 21 CFR standards, these shatterproof bottles offer a safer, high-impact alternative to glass in high-handling environments. The Narrow Mouth LDPE Reagent Bottle design ensures precision dispensing, controlled pouring, and minimal spillage. LDPE chemical resistant reagent bottles offer excel chemical resistance and have high impact strength
Features
LDPE storage reagent bottles are flexible and squeezable for easy dispensing.
LDPE storage bottles are lightweight and crack-resistant.
LDPE Bottles with low Liquid Retention
Leak-proof Linerless screw caps for secure sealing.