Learn More
Ace Glass, Inc. 500mL separatory funnel with locking stopper
SDP
Catalog No. 50974700
Encompass_Preferred
500ml separatory funnel with locking stopper that will not fall out during shaking or transport. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.
Catalog No. 50-974-700 Supplier Ace Glass, Inc. Supplier No. 723161
500ml separatory funnel with locking stopper that will not fall out during shaking or transport. Stopper can be secured in top joint via cap and O-ring for a leak-tight seal.