Learn More
Chemglass Life Sciences Bottle, Media, 500mL, Square, PET, Sterile, 8/Pk, 24/Case
SDP
Catalog No. 501897805
Encompass_Preferred
Break-resistant polyethylene terephthalate (PET) with transparency equivalent to glass.High-density Polyethylene (HDPE) cap.Smooth surface, not easy to residue, easy to clean.Good chemical resistance, suitable for sample collection of various liquid particle size test, high clean reagents, particle standard materials and standard samples.With scale, white enamel marking area, durable.Excellent physical and mechanical properties over a wide temperature range.The bottle body is square, easy to hold, easy to store.Unique external thick collar design for easy sealing operation in experiments.Sterile, DNase/RNase/Pyrogen free
Catalog No. 50-189-7805 Supplier Chemglass Life Sciences Supplier No. CGN1151500
Break-resistant polyethylene terephthalate (PET) with transparency equivalent to glass.High-density Polyethylene (HDPE) cap.Smooth surface, not easy to residue, easy to clean.Good chemical resistance, suitable for sample collection of various liquid particle size test, high clean reagents, particle standard materials and standard samples.With scale, white enamel marking area, durable.Excellent physical and mechanical properties over a wide temperature range.The bottle body is square, easy to hold, easy to store.Unique external thick collar design for easy sealing operation in experiments.Sterile, DNase/RNase/Pyrogen free